Triisopropanolamine, 95%
Triisopropanolamine, 95%
Triisopropanolamine, 95%
Triisopropanolamine, 95%
Thermo Scientific Chemicals
Triisopropanolamine, 95%
CAS: 122-20-3 | C9H21NO3 | 191.271 g/mol
Have Questions?
Catalog NumberQuantity
ALFA19966.36500 g
Catalog number ALFA19966.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTriisopropanolamine
CAS122-20-3
Health Hazard 1H319
Health Hazard 2GHS H Statement
H319
Causes serious eye irritation.
Health Hazard 3P264b-P280i-P305+P351+P338
View more
Major applications include water-based coating applications and agricultural products. Additional applications are antistat agents for polymers, corrosion inhibitor, electrodeposition/electrocoating, lubricants, paper, pigment dispersion, plastics, polyurethane additive, reaction intermediates, rubber curing, surfactants, mineral dispersion, and urethanes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Major applications include water-based coating applications and agricultural products. Additional applications are antistat agents for polymers, corrosion inhibitor, electrodeposition/electrocoating, lubricants, paper, pigment dispersion, plastics, polyurethane additive, reaction intermediates, rubber curing, surfactants, mineral dispersion, and urethanes.
Solubility
Insoluble in water. Soluble ethanol, diethyl ether, chloroform (slightly ), and methanol (>500 g/100g).
Notes
Hygroscopic. Store away from oxidizing agents, water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only
General References:
  1. Pensri Piboonchaisit, et al. Novel Route to Tris(silatranyloxy-i-propyl)amine Directly from Silica and Triisopropanolamine, Part I.ScienceAsia.1999,25113-119.
  2. Paul J. Sandberg.; F. Doncaster. On the mechanism of strength enhancement of cement paste and mortar with triisopropanolamine.Cem. Concr. Res.2004,34(6), 973-976.