Zinc selenide
Zinc selenide
Zinc selenide
Thermo Scientific Chemicals
Zinc selenide
CAS: 1315-09-9 | SeZn | 144.35 g/mol
Have Questions?
Catalog NumberQuantity
ALFA19947.30250 g
Catalog number ALFA19947.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialZinc selenide
Melting Point>1100°C
CAS1315-09-9
Health Hazard 1H301-H373
Health Hazard 2GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
View more
Zinc selenide is used to form II-VI light-emitting diodes and diode lasers. It is doped with chromium (ZnSe:Cr) has been used as an infrared laser gain medium. It is used as an infrared optical material. It is used in x-ray and gamma ray detectors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Zinc selenide is used to form II-VI light-emitting diodes and diode lasers. It is doped with chromium (ZnSe:Cr) has been used as an infrared laser gain medium. It is used as an infrared optical material. It is used in x-ray and gamma ray detectors.
Solubility
Insoluble in water.
Notes
Incompatible with oxidizing agents and bases. Store in a cool, dry conditions in a well sealed container.
RUO – Research Use Only
General References:
  1. Evgeni Sorokin; Nikolai Tolstik; Kathleen I Schaffers; Irina T Sorokina. Femtosecond SESAM-modelocked Cr:ZnS laser. Optics Express. 2012, 20, (27), 28947-28952.
  2. Damian W Synnott; Michael K Seery; Steven J Hinder; John Colreavy; Suresh C Pillai. Novel microwave assisted synthesis of ZnS nanomaterials. Nanotechnology. 2013, 24, (4), 045704.