1-Tetradecanol, 97+%
1-Tetradecanol, 97+%
1-Tetradecanol, 97+%
1-Tetradecanol, 97+%
Thermo Scientific Chemicals
1-Tetradecanol, 97+%
CAS: 112-72-1 | C14H30O | 214.39 g/mol
Have Questions?
Catalog NumberQuantity
ALFA19638.36500 g
Catalog number ALFA19638.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Tetradecanol
CAS112-72-1
Health Hazard 1H319
Health Hazard 2GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P264b-P280i-P305+P351+P338
View more
1-Tetradecanol is used as an ingredient in cosmetics such as cold creams. It is an active intermediate in the chemical synthesis of sulfated alcohol. It is also employed in the fabrication of temperature-regulated drug release system based on phase-change materials. It plays a vital role in filling the hollow interiors of gold nanocages in the fabrication of new theranostic system, which has unique feature of photoacoustic imaging.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Tetradecanol is used as an ingredient in cosmetics such as cold creams. It is an active intermediate in the chemical synthesis of sulfated alcohol. It is also employed in the fabrication of temperature-regulated drug release system based on phase-change materials. It plays a vital role in filling the hollow interiors of gold nanocages in the fabrication of new theranostic system, which has unique feature of photoacoustic imaging.
Solubility
Insoluble in water. Soluble in diethyl ether. Slightly soluble in ethanol.
Notes
Incompatible with strong oxidizing agents, acid chlorides and acid anhydrides. Store in a cool place.
RUO – Research Use Only
General References:
  1. Tian, T.; Song, J.; Niu, L.; Feng, R. Preparation and properties of 1-tetradecanol/1,3:2,4-di-(3,4-dimethyl) benzylidene sorbitol gelatinous form-stable phase change materials. Thermochim. Acta 2013, 554, 54-58.
  2. Corberan, V. C.; Gomez-Aviles, A.; Martinez-Gonzalez, S.; Ivanova, S.; Dominguez, M. I.; Gonzalez-Perez, M. E. Heterogeneous selective oxidation of fatty alcohols: Oxidation of 1-tetradecanol as a modelsubstrate. Catal. Today 2014, 238, 49-53.