N,N'-Diisopropylcarbodiimide, 99%
N,N'-Diisopropylcarbodiimide, 99%
N,N'-Diisopropylcarbodiimide, 99%
N,N'-Diisopropylcarbodiimide, 99%
Thermo Scientific Chemicals
N,N'-Diisopropylcarbodiimide, 99%
Alfa Aesar N,N'-Diisopropylcarbodiimide, 99%, Quantity: 25g, Boiling Point: 145 deg.C to 148 deg.C, Molecular Weight: 126.203, Percent Purity: 99%, Beilstein: 878281, CAS: 693-13-0, ChEBI: CHEBI:53092, Density: 0.81, DOT Information: Hazard Class: 6.1; Packaging Group: I, EINECS Number: 211-743-7 | CAS: 693-13-0 | C7H14N2 | 126.203 g/mol
Have Questions?
Catalog NumberQuantity
ALFA19292.0910 g
Catalog number ALFA19292.09
Price (MYR)
459.00
EA
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (MYR)
459.00
EA
Add to cart
Specifications
Chemical Name or MaterialN,N'-Diisopropylcarbodiimide
CAS693-13-0
Health Hazard 1H226-H315-H317-H318-H330-H334-H335
Health Hazard 2GHS H Statement
H330-H334-H318-H226-H315-H317-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Flammable liquid and vapour.
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P272-P280-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P333+P313-P363-P370+P378q-P501c
View more
N,N'-Diisopropylcarbodiimide is used as a reagent in synthetic organic chemistry. It serves as a chemical intermediate and as a stabilizer for Sarin (chemical weapon). It is also used in the synthesis of peptide and nucleic acid. Further, it is used as an antineoplastic and involved in the treatment of malignant melanoma and sarcomas. In addition to this, it is used in the synthesis of acid anhydride, aldehyde, ketone and isocyanate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N,N′-Diisopropylcarbodiimide is used as a reagent in synthetic organic chemistry. It serves as a chemical intermediate and as a stabilizer for Sarin (chemical weapon). It is also used in the synthesis of peptide and nucleic acid. Further, it is used as an antineoplastic and involved in the treatment of malignant melanoma and sarcomas. In addition to this, it is used in the synthesis of acid anhydride, aldehyde, ketone and isocyanate.
Solubility
Soluble in chloroform, methylene chloride, acetonitrile, dioxane, dimethylformamide andtetrahydrofuran.
Notes
Moisture sensitive. Store in a cool place Incompatible with oxidizing agents and acids.
RUO – Research Use Only
General References:
  1. Reagent for peptide coupling: J. Org. Chem., 44, 5000 (1979). Has the advantage over N,N'-Dicyclohexyl carbodiimide, A10973 in solid phase peptide syntheses that the urea by-product is more soluble in organic solvents and hence more readily separated from the polymer support. For peptide reagents, see Appendix 6.
  2. Ogawa, T.; Usuki, N.; Nakazono, K.; Koyama, Y.; Takata, T. Linear-cyclic polymer structural transformation and its reversible control using a rational rotaxane strategy. Chem. Commun. 2015, 51 (26), 5606-5609.
  3. Smith, J. M.; Moreno, J.; Boal, B. W.; Garg, N. K. Cascade Reactions: A Driving Force in Akuammiline Alkaloid Total Synthesis. Angew. Chem. Int. Ed. 2015, 54 (2), 400-412.