4-Nitroanisole, 97%
4-Nitroanisole, 97%
4-Nitroanisole, 97%
Thermo Scientific Chemicals
4-Nitroanisole, 97%
CAS: 100-17-4 | C7H7NO3 | 153.14 g/mol
Have Questions?
Catalog NumberQuantity
ALFA19152.30250 g
Catalog number ALFA19152.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Nitroanisole
CAS100-17-4
Health Hazard 1H341
Health Hazard 2GHS H Statement
H341
Suspected of causing genetic defects.
Health Hazard 3P201-P202-P281-P308+P313-P501c