4,4'-Diaminodiphenyl sulfone, 98%
4,4'-Diaminodiphenyl sulfone, 98%
4,4'-Diaminodiphenyl sulfone, 98%
4,4'-Diaminodiphenyl sulfone, 98%
Thermo Scientific Chemicals
4,4'-Diaminodiphenyl sulfone, 98%
CAS: 80-08-0 | C12H12N2O2S | 248.3 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFA18976.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or Material4,4'-Diaminodiphenyl sulfone
CAS80-08-0
Health Hazard 1H302-H373
Health Hazard 2GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3P260-P264b-P270-P301+P312-P314-P330-P501c
View more
It is active against a wide range of bacteria, but is mainly employed for its actions against Mycobacterium leprae, being used as part of multidrug regimens in the treatment of all forms of leprosy. It is a second-line medication for the treatment and prevention of Pneumocystis pneumonia and for the prevention of toxoplasmosis in those who have poor immune function. Additionally, it has been used for acne as well as other skin conditions. Moreover it is employed as an anti-inflammatory drug, anti-infective drug, a leprostatic drug and an antimalarial drug.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Insoluble in water. Solubility in hot methanol is almost transparent. Soluble in DMSO, ethanol and acetone.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. Patel AP, Patil AS. Dapsone for immune thrombocytopenic purpura in children and adults.Therapeutic drug monitoring.2014,36(2), 261-263.
  2. B.L. Maloff, et al. Dapsone inhibits LTB4 binding and bioresponse at the cellular and physiologic levels.Eur. J. Pharmacol.1988158(1-2), 85-89.