Sodium cyclamate, 98%
Sodium cyclamate, 98%
Sodium cyclamate, 98%
Sodium cyclamate, 98%
Thermo Scientific Chemicals
Sodium cyclamate, 98%
CAS: 139-05-9 | C6H12NNaO3S | 201.22 g/mol
Have Questions?
Catalog NumberQuantity
ALFA18666.30250 g
Catalog number ALFA18666.30
Price (MYR)
432.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
432.00
EA
Add to cart
Specifications
Chemical Name or MaterialSodium cyclamate
CAS139-05-9
Health Hazard 1H302
Health Hazard 3P264b-P270-P301+P312-P330-P501c
Melting Point>300°C
View more
Sodium Cyclamate is an artificial sweetener and is 30 times as saccharose's. It is widely used in pickles, seasoning sauce, cakes, biscuits, bread, ice cream, frozen sucker, popsicles, drinks and so on, with a maximum amount of 0.65g/kg. Secondly, it is used in confect, with a maximum amount of 1.0g/kg. Thirdly, it is used in orange peel, preserved plum, dried arbutus and so on, with the largest amount of 8.0g/kg. It is also used in cosmetics and pharmaceutical industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium Cyclamate is an artificial sweetener and is 30 times as saccharose′s. It is widely used in pickles, seasoning sauce, cakes, biscuits, bread, ice cream, frozen sucker, popsicles, drinks and so on, with a maximum amount of 0.65g/kg. Secondly, it is used in confect, with a maximum amount of 1.0g/kg. Thirdly, it is used in orange peel, preserved plum, dried arbutus and so on, with the largest amount of 8.0g/kg. It is also used in cosmetics and pharmaceutical industry.
Solubility
Soluble in water (>=10 g/100 mL at 20°C).
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. F.J.C. Roe.; L.S. Levy.; R.L. Carter. Feeding studies on sodium cyclamate, saccharin and sucrose for carcinogenic and tumour-promoting activity.Food and Cosmetics Toxicology,1970,8(2), 135-145.
  2. Thomas A. Biemer. Analysis of saccharin, acesulfame-K and sodium cyclamate by high-performance ion chromatography.J. Chromatogr. A,1989,463463-468.