Copper(II) formate tetrahydrate, 98%
Copper(II) formate tetrahydrate, 98%
Copper(II) formate tetrahydrate, 98%
Copper(II) formate tetrahydrate, 98%
Thermo Scientific Chemicals
Copper(II) formate tetrahydrate, 98%
CAS: 5893-61-8 | C2H10CuO8 | 225.64 g/mol
Have Questions?
Catalog NumberQuantity
ALFA18569.1850 g
Catalog number ALFA18569.18
Price (MYR)
422.00
EA
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
422.00
EA
Add to cart
Specifications
Chemical Name or MaterialCopper(II) formate tetrahydrate
CAS5893-61-8
Health Hazard 1H302-H315-H318-H335
Health Hazard 2GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
View more
It is employed as an antibacterial agent in the treatment of cellulose.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as an antibacterial agent in the treatment of cellulose.
Solubility
Soluble in water (125 g/L at 20°C). Very slightly soluble in alcohol. Practically insoluble in most organic solvents.
Notes
Store away from oxidizing agents and strong bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. Hanako Kobayashi and Taiichiro Haseda. The Magnetic Properties of Cupric Formate Tetrahydrate at Low Temperatures.J. Phys. Soc. Jpn.1963,18541-550.
  2. Takasuke Matsuo.; Yoshio Kume.; Hiroshi Suga.; SyûzÔ Seki. Heat capacities of copper(II) formate tetrahydrate and tetradeuterate: A comparative study of phase transitions in layer hydrate crystals.J. Phys. Chem. Solids,1976,37(5), 499-506.