1,4-Bis(trifluoromethyl)benzene, 99%
1,4-Bis(trifluoromethyl)benzene, 99%
1,4-Bis(trifluoromethyl)benzene, 99%
1,4-Bis(trifluoromethyl)benzene, 99%
Thermo Scientific Chemicals
1,4-Bis(trifluoromethyl)benzene, 99%
CAS: 433-19-2 | C8H4F6 | 214.11 g/mol
Have Questions?
Catalog NumberQuantity
ALFA18411.1425 g
Catalog number ALFA18411.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material1,4-Bis(trifluoromethyl)benzene
CAS433-19-2
Health Hazard 1Danger
Health Hazard 2GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
P210-P261-P280g-P305+P351+P338
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.