N,N-Dimethyl-4-nitrosoaniline, 98%
N,N-Dimethyl-4-nitrosoaniline, 98%
N,N-Dimethyl-4-nitrosoaniline, 98%
N,N-Dimethyl-4-nitrosoaniline, 98%
Thermo Scientific Chemicals
N,N-Dimethyl-4-nitrosoaniline, 98%
CAS: 138-89-6 | C8H10N2O | 150.181 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
25 g
100 g
Catalog number ALFA18301.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialN,N-Dimethyl-4-nitrosoaniline
CAS138-89-6
Health Hazard 1H301+H311+H331-H373-H500-X
Health Hazard 2GHS H Statement
H228-H251-H301-H310-H351-H373
Flammable solid.
Self-heating: may catch fire.
Toxic if swallowed.
Fatal in contact with skin.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3P210-P235+P410-P260-P262-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P362-P370+P380-P380-P501c