meso-Tetraphenylporphine, low chlorine
meso-Tetraphenylporphine, low chlorine
meso-Tetraphenylporphine, low chlorine
meso-Tetraphenylporphine, low chlorine
Thermo Scientific Chemicals
meso-Tetraphenylporphine, low chlorine
CAS: 917-23-7 | C44H30N4 | 614.75 g/mol
Have Questions?
Catalog NumberQuantity
ALFA18261.031 g
Catalog number ALFA18261.03
Price (MYR)
437.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
437.00
EA
Add to cart
Specifications
Chemical Name or Materialmeso-Tetraphenylporphine
Name Notelow chlorine
CAS917-23-7
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
meso-Tetraphenylporphyrin is a porphyrin-based compound of interest in molecular electronics and supramolecular building blocks. Porphyrins are often used to construct structures in supramolecular chemistry. It finds it application as a sensitizer used for the photochemical generation of singlet oxygen.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
meso-Tetraphenylporphyrin is a porphyrin-based compound of interest in molecular electronics and supramolecular building blocks. Porphyrins are often used to construct structures in supramolecular chemistry. It finds it application as a sensitizer used for the photochemical generation of singlet oxygen.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. Hans-Heinrich Limbach.; Jürgen Hennig.; and Josef Stulz. IR-spectroscopic study of isotope effects on the NHD-stretching bands of meso-tetraphenylporphine and vibrational hydrogen tunneling.J. Chem. Phys.1983,78(9), 5432.
  2. Alan D. Adler.; Frederick R. Longo.; John D. Finarelli.; Joel Goldmacher.; Jacques Assour.; Leonard Korsakoff. A simplified synthesis for meso-tetraphenylporphine.J. Org. Chem.,1967,32(2), 476-476.