CHES, 99%
CHES, 99%
CHES, 99%
Thermo Scientific Chemicals
CHES, 99%
CAS: 103-47-9 | C8H17NO3S | 207.288 g/mol
Have Questions?
Catalog NumberQuantity
ALFA18047.1425 g
Catalog number ALFA18047.14
Price (MYR)
453.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
453.00
EA
Add to cart
Specifications
Chemical Name or MaterialCHES
CAS103-47-9
Melting Point>300°C
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H319
Causes serious eye irritation.
View more
CHES is zwitterionic buffer useful in pH range 8.6 to 10.0.CHES is widely utilized as a buffer which is used for investigations on pH-dependent processes in enzymology. It has been shown to have an unusually high affinity for the iodoacetate binding site of liver alcohol dehydrogenase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
CHES is zwitterionic buffer useful in pH range 8.6 to 10.0.CHES is widely utilized as a buffer which is used for investigations on pH-dependent processes in enzymology. It has been shown to have an unusually high affinity for the iodoacetate binding site of liver alcohol dehydrogenase.
Solubility
Soluble in water.
Notes
Store in cool place. Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Biological buffer, pKa = 9.3 at 20°: Biochemistry, 5, 467 (1966).
  2. Ferreira, C. M. H.; Pinto, I. S. S.; Soares, E. V.; Soares, H. M. V. M. (Un)suitability of the use of pH buffers in biological, biochemical and environmental studies and their interaction with metal ions - a review. RSC Adv. 2015, 5 (39), 30989-31003.
  3. Devi, T. S. R.; kumar, J. S.; Ramkumaar, G. R. DFT analysis on the molecular structure, vibrational and electronic spectra of 2-(cyclohexylamino)ethanesulfonic acid. Spectrochim. Acta, Part A 2015, 137, 761-777.