Lithium acetate dihydrate, 99%
Lithium acetate dihydrate, 99%
Lithium acetate dihydrate, 99%
Lithium acetate dihydrate, 99%
Lithium acetate dihydrate, 99%
Thermo Scientific Chemicals
Lithium acetate dihydrate, 99%
CAS: 6108-17-4 | C2H7LiO4 | 102.014 g/mol
Have Questions?
Catalog NumberQuantity
ALFA17921.30250 g
Catalog number ALFA17921.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialLithium acetate dihydrate
CAS6108-17-4
Health Hazard 1H319
Health Hazard 3P264b-P280i-P305+P351+P338
Melting Point54°C to 57°C
View more
Lithium acetate dihydrate is used as a buffer component in the modified ninhydrin reagent for amino analysis by ion-exchange chromatography. It is used as a catalyst in organic reactions, alkyd resin and acrylic polymer production and as an anticorrosion agent in molding polyphenylene sulfide resins as well as in the separation of saturated and unsaturated fatty acids. In the pharmaceutical industry, it is utilized for the preparation of diuretics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Lithium acetate dihydrate is used as a buffer component in the modified ninhydrin reagent for amino analysis by ion-exchange chromatography. It is used as a catalyst in organic reactions, alkyd resin and acrylic polymer production and as an anticorrosion agent in molding polyphenylene sulfide resins as well as in the separation of saturated and unsaturated fatty acids. In the pharmaceutical industry, it is utilized for the preparation of diuretics.
Solubility
Soluble in water and ethanol.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Schroder, F.; Winkler, B.; Bauer, J. D.; Haussuhl, E.; Escoto, B. R.; Lopez, F. T.; Borja, M. A.; Richter, C.; Ferner, J. Evidence for low-temperature isotopic ordering in methyl-monodeuterated lithium acetate dihydrate. J. Chem. Thermodyn. 2012, 54, 373-376.
  2. Winkler, B.; Haussuhl, E.; Bauer, J. D.; Schroder, F.; Refson, K.; Milman, V.; Hennion, B.; Bossak, A. Krisch, M. Influence of deuteration on lithium acetate dihydrate studied by inelastic X-ray scattering, density functional theory, thermal expansion, elastic and thermodynamic measurements. Dalton Trans. 2011, 40 (8), 1737-1742.