Wright's Stain
Wright's Stain
Wright's Stain
Thermo Scientific Chemicals
Wright's Stain
CAS: 68988-92-1 | C36H27Br4N3O5S+2 | 933.304 g/mol
Have Questions?
Catalog NumberQuantity
ALFA17903.1425 g
Catalog number ALFA17903.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialWright's Stain
CAS68988-92-1
Health Hazard 1H302-H318
Health Hazard 2GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
Health Hazard 3P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
View more
Useful stain for blood and for bone marrow films