DL-Malic acid, 98%
DL-Malic acid, 98%
DL-Malic acid, 98%
Thermo Scientific Chemicals
DL-Malic acid, 98%
CAS: 6915-15-7 | C4H6O5 | 134.09 g/mol
Have Questions?
Catalog NumberQuantity
ALFA17874.30250 g
Catalog number ALFA17874.30
Price (MYR)
267.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
267.00
EA
Add to cart
Specifications
Chemical Name or MaterialDL-Malic acid
CAS6915-15-7
Melting Point130°C to 133°C
Health Hazard 1H319
Health Hazard 3P264b-P280i-P305+P351+P338
View more
DL-Malic acid is used as a food additive. It is a component of some artificial vinegar flavors. Further, it is used in the preparation of chiral compounds, including κ-opioid rece. It is utilized for chiral resolution by ligand-exchange capillary electrophoresis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
DL-Malic acid is used as a food additive. It is a component of some artificial vinegar flavors. Further, it is used in the preparation of chiral compounds, including κ-opioid rece. It is utilized for chiral resolution by ligand-exchange capillary electrophoresis.
Solubility
Soluble in acetone, ether, water, methanol and ethanol.
Notes
Incompatible with bases, oxidizing agents, reducing agents and alkali metals.
RUO – Research Use Only