DL-Glutamic acid monohydrate, 98+%
DL-Glutamic acid monohydrate, 98+%
DL-Glutamic acid monohydrate, 98+%
DL-Glutamic acid monohydrate, 98+%
Thermo Scientific Chemicals
DL-Glutamic acid monohydrate, 98+%
CAS: 19285-83-7 | C5H11NO5 | 165.145 g/mol
Have Questions?
Catalog NumberQuantity
ALFA17719.1425 g
Catalog number ALFA17719.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDL-Glutamic acid monohydrate
Melting Point∼200°C (decomposition)
CAS19285-83-7
Recommended StorageAmbient temperatures
RTECS NumberLZ9648000