D-Tyrosine, 99%
D-Tyrosine, 99%
D-Tyrosine, 99%
Thermo Scientific Chemicals
D-Tyrosine, 99%
CAS: 556-02-5 | C9H11NO3 | 181.19 g/mol
Have Questions?
Catalog NumberQuantity
ALFA17709.031 g
Catalog number ALFA17709.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialD-Tyrosine
Melting Point310°C to 314°C
CAS556-02-5
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
D-Tyrosine is synthesized from phenylalanine. It is also the precursor of epinephrine, thyroid hormones, and melanin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
D-Tyrosine is synthesized from phenylalanine. It is also the precursor of epinephrine, thyroid hormones, and melanin.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
  1. Victor J. Hruby.; Donald A. Upson.; Diane M. Yamamoto.; Clark W. Smith.; Roderich Walter. Active site studies of neurohypophyseal hormones. Comparison of oxytocin and arginine vasopressin analogs containing 2-D-tyrosine. J. Am. Chem. Soc. 1979, 101, (10), 2717-2721.
  2. Richard Calendar.; Paul Berg. d-Tyrosyl RNA: Formation, hydrolysis and utilization for protein synthesis. Journal of Molecular Biology. 1967, 26, (1), 39-54.