Polyethylene glycol 6,000
Polyethylene glycol 6,000
Polyethylene glycol 6,000
Polyethylene glycol 6,000
Thermo Scientific Chemicals
Polyethylene glycol 6,000
CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number ALFA17541.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPolyethylene Glycol 6000
CAS25322-68-3
Physical FormPowder
Health Hazard 1H315-H320
Health Hazard 3P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P501c
View more
Polyethylene glycol is used in the modification of therapeutic proteins and peptides to enhance its solubility. It is also used in the fabrication of bioactive and immuno isolating barriers for encapsulation of cells. In Gas chromatorgraphy, it is used as a polar stationary phase. It is used as mold release agent and lubricant in fabricating elastomers.