DL-Isoleucine, 99%
DL-Isoleucine, 99%
DL-Isoleucine, 99%
Thermo Scientific Chemicals
DL-Isoleucine, 99%
CAS: 443-79-8 | C6H13NO2 | 131.18 g/mol
Have Questions?
Catalog NumberQuantity
ALFA17521.22100 g
Catalog number ALFA17521.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDL-Isoleucine
Melting Point∼280°C (decomposition)
CAS443-79-8
Recommended StorageAmbient temperatures
EINECS Number207-139-8