Azure I
Azure I
Azure I
Thermo Scientific Chemicals
Azure I
CAS: 531-55-5 | C15H16ClN3S | 305.824 g/mol
Have Questions?
Catalog NumberQuantity
ALFA17508.065 g
Catalog number ALFA17508.06
Price (MYR)
438.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
438.00
EA
Add to cart
Specifications
Chemical Name or MaterialAzure I
CAS531-55-5
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H341
Suspected of causing genetic defects.
Health Hazard 3GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
View more
Azure B is used for staining semi-thin sections of plant tissue. This dye is also useful in the NCCLS method for blood smears and Lillie's modified Nocht's method for paraffin sections.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Azure B is used for staining semi-thin sections of plant tissue. This dye is also useful in the NCCLS method for blood smears and Lillie′s modified Nocht′s method for paraffin sections.
Solubility
Soluble in 1 mg/ml water. Sparingly soluble in alcohol.
Notes
Store in a cool place. Ensure proper ventilation. Keep away from oxidizing agents.
RUO – Research Use Only
General References:
  1. Chen-Xin Cai.; Kuan-Hong Xue. Electrocatalysis of NADH oxidation with electropolymerized films of azure I. J. Electroanal. Chem. 1997, 427 (1-2), 147-153.
  2. Na Li .; Ruo Yuan.; Yaqin Chai.; Shihong Chen.; Haizhen An.; Wenjuan Li. New Antibody Immobilization Strategy Based on Gold Nanoparticles and Azure I/Multi-Walled Carbon Nanotube Composite Membranes for an Amperometric Enzyme Immunosensor. J. Phys. Chem. 2007, 111 (24),8443-8450.