Sodium chromate tetrahydrate, 99+%
Sodium chromate tetrahydrate, 99+%
Sodium chromate tetrahydrate, 99+%
Sodium chromate tetrahydrate, 99+%
Thermo Scientific Chemicals
Sodium chromate tetrahydrate, 99+%
CAS: 10034-82-9 | CrH8Na2O8 | 234.032 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFA17499.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium chromate tetrahydrate
CAS10034-82-9
Health Hazard 1H301-H312-H314-H317-H330-H334-H335-H340-H350-H360-H372
Health Hazard 2GHS H Statement
H301-H330-H334-H340-H350-H360-H372-H314-H318-H312-H317
Toxic if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
View more
Sodium chromate tetrahydrate is used as a corrosion inhibitor, wood preservative and an auxiliary in textile industry. It serves as an oxidant, which is used to prepare carboxylic acids and ketones from primary and secondary alcohols respectively. Further, it is used as a starting material to prepare other chromium compounds and as a catalyst in organic synthesis especially pharmaceuticals and agro chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium chromate tetrahydrate is used as a corrosion inhibitor, wood preservative and an auxiliary in textile industry. It serves as an oxidant, which is used to prepare carboxylic acids and ketones from primary and secondary alcohols respectively. Further, it is used as a starting material to prepare other chromium compounds and as a catalyst in organic synthesis especially pharmaceuticals and agro chemicals.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible withstrong reducing agents, organic materials, strong acids, combustible materials and powdered metals.
RUO – Research Use Only
General References:
  1. King, S.; Benson, S.; Kelly, T.; Lennard, C. Determining the effects of routine fingermark detection techniques on the subsequent recovery and analysis of explosive residues on various substrates. Forensic Sci. Int. 2013, 233 (1), 257-264.
  2. Ayarza, N.; Góngora, J. M.; Alonso, R. M.; Jiménez, R. M. Determination of anionic impurities in hexafluorosilicic acid by capillary zone electrophoresis. Anal. Methods 2012, 4 (10), 3157-3162.