Creatine, anhydrous, 98%
Creatine, anhydrous, 98%
Creatine, anhydrous, 98%
Creatine, anhydrous, 98%
Thermo Scientific Chemicals
Creatine, anhydrous, 98%
CAS: 57-00-1 | C4H9N3O2 | 131.135 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFA17477.22
Price (MYR)
816.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
816.00
EA
Add to cart
Specifications
Chemical Name or MaterialCreatine, Anhydrous
Name Noteanhydrous
CAS57-00-1
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H315-H319-H335
View more
Creatine is involved in the rapid production of ATP associated with creatine kinases in skeletal muscle tissue. Creatine may be used as a supplement to study its mechanism of action. It acts as a nutraceutical and a neuroprotective agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Creatine is involved in the rapid production of ATP associated with creatine kinases in skeletal muscle tissue. Creatine may be used as a supplement to study its mechanism of action. It acts as a nutraceutical and a neuroprotective agent.
Solubility
Soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. Persky AM, et al. Clinical pharmacology of the dietary supplement creatine monohydrate.Pharmacological reviews.,2001,53(2), 161-176.
  2. Mendes RR, et al. [Creatine: the nutritional supplement for exercise - current concepts].Archivos latinoamericanos de nutricion.,2002,52(2), 117-127.