3,3',5,5'-Tetrabromobisphenol A, 97%
3,3',5,5'-Tetrabromobisphenol A, 97%
3,3',5,5'-Tetrabromobisphenol A, 97%
3,3',5,5'-Tetrabromobisphenol A, 97%
Thermo Scientific Chemicals
3,3',5,5'-Tetrabromobisphenol A, 97%
CAS: 79-94-7 | C15H12Br4O2 | 543.875 g/mol
Have Questions?
Catalog NumberQuantity
ALFA17395.22100 g
Catalog number ALFA17395.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material3,3',5,5'-Tetrabromobisphenol A
CAS79-94-7
Melting Point179°C to 182°C
Recommended StorageAmbient temperatures
DOT InformationTransport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.