4-(Chlorosulfonyl)benzoic acid, 96%
4-(Chlorosulfonyl)benzoic acid, 96%
4-(Chlorosulfonyl)benzoic acid, 96%
4-(Chlorosulfonyl)benzoic acid, 96%
Thermo Scientific Chemicals
4-(Chlorosulfonyl)benzoic acid, 96%
CAS: 10130-89-9 | C7H5ClO4S | 220.623 g/mol
Have Questions?
Catalog NumberQuantity
ALFA17337.065 g
Catalog number ALFA17337.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material4-(Chlorosulfonyl)benzoic acid
CAS10130-89-9
Health Hazard 1H302-H314-H335
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
View more
It is a chemical reagent used in catalyzing living radical polymerizations, in the preparation of polymer bound transfer hydrogenation catalyst1 and 4-(carboxymethyl-sulfamoyl)-benzoyladenosine. It is also used as a reagent in the sulfonation of γ-cyclodextrin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a chemical reagent used in catalyzing living radical polymerizations, in the preparation of polymer bound transfer hydrogenation catalyst1 and 4-(carboxymethyl-sulfamoyl)-benzoyladenosine. It is also used as a reagent in the sulfonation of γ-cyclodextrin.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only
General References:
  1. Bayston DJ, et al. Synthesis and evaluation of a chiral heterogeneous transfer hydrogenation catalyst.Tetrahedron Asymmetry.,1988,9(12), 2015-18.
  2. Nguyen-Hai Nam, et al. ATP-phosphopeptide conjugates as inhibitors of Src tyrosine kinases.Bioorg. Med. Chem.,2004,12(22), 5753-5766 .