Nile Blue A
Nile Blue A
Nile Blue A
Thermo Scientific Chemicals
Nile Blue A
CAS: 3625-57-8 | C40H40N6O6S | 732.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFA17174.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialNile Blue A
Melting Point>300°C (decomposition)
CAS3625-57-8
Recommended StorageAmbient temperatures
RTECS NumberDJ1931000
View more
Used in the preparation of an amperometric glucose sensor. Also can be employed in Lillie and Fullmer method for staining and discriminating between melanins and lipofuscins in paraffin sections of animal tissue; Lillie and Fuller method for fatty acids/lipids using frozen sections of fatty liver and avocado.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used in the preparation of an amperometric glucose sensor. Also can be employed in Lillie and Fullmer method for staining and discriminating between melanins and lipofuscins in paraffin sections of animal tissue; Lillie and Fuller method for fatty acids/lipids using frozen sections of fatty liver and avocado.
Solubility
Soluble in ethanol, and water (50 g/L) at 25°C.
Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
RUO – Research Use Only
General References:
  1. Jason D Slinker.; Natalie B Muren.; Sara E Renfrew.; Jacqueline K Barton. DNA charge transport over 34 nm.Nat Chem.2011,3(3), 228-233.
  2. Meikun Fan.; Penghui Wang.; Carlos Escobedo.; David Sinton.; Alexandre G Brolo. Surface-enhanced Raman scattering (SERS) optrodes for multiplexed on-chip sensing of nile blue A and oxazine 720.Lab Chip.2012,12,(8), 1554-1560 .