o-Dianisidine, 98+%
o-Dianisidine, 98+%
o-Dianisidine, 98+%
Thermo Scientific Chemicals
o-Dianisidine, 98+%
CAS: 119-90-4 | C14H16N2O2 | 244.294 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFA17150.14
Price (MYR)
744.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
744.00
EA
Add to cart
Specifications
Chemical Name or Materialo-Dianisidine
CAS119-90-4
Health Hazard 1H302-H350
Health Hazard 2GHS H Statement
H350-H302
Health Hazard 3P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
View more
o-Dianisidine is used as a redox indicator. It is used as an intermediate for the preparation of azo dyes, pigments and o-dianisidine diisocyanate, which finds application in adhesives, polyurethane elastomers and resins. It is also used in various industries like leather, paper, plastics, rubber, and textiles. It acts as a reagent to detect metals, thiocyanates, and nitrites.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
o-Dianisidine is used as a redox indicator. It is used as an intermediate for the preparation of azo dyes, pigments and o-dianisidine diisocyanate, which finds application in adhesives, polyurethane elastomers and resins. It is also used in various industries like leather, paper, plastics, rubber, and textiles. It acts as a reagent to detect metals, thiocyanates, and nitrites.
Solubility
Soluble in alcohol, benzene, ether, chloroform, acetone, most organic solvents and lipids. Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Redox indicator.
  2. Perju, E.; Cozan, V.; Marin, L.; Bruma, M. Semiflexible thermotropic polyazomethines based on o-dianisidine mesogenic core. Liq. Cryst. 2015, 42 (9), 1309-1319.
  3. Sundaram, S.; Jagannathan, M.; Kadir, M. R. A.; Palanivel, S.; Hadibarata, T.; Yusoff, A. R. M. A new electro-generated o-dianisidine derivative stabilized MWCNT-modified GCE for low potential gallic acid detection. RSC Adv. 2015, 5 (57), 45996-46006.