Phenolphthalein, 98%
Phenolphthalein, 98%
Phenolphthalein, 98%
Phenolphthalein, 98%
Thermo Scientific Chemicals
Phenolphthalein, 98%
CAS: 77-09-8 | C20H14O4 | 318.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number ALFA17135.30
Price (MYR)
419.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
419.00
EA
Add to cart
Specifications
Chemical Name or MaterialPhenolphthalein
CAS77-09-8
Health Hazard 1H341-H351-H361f-H373
Health Hazard 2GHS H Statement
H350-H341-H361
May cause cancer.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
Health Hazard 3P201-P202-P260-P281-P308+P313-P501c
View more
Phenolphthalein is widely used as an indicator in acid-base titrations, water treatment applications. Due to its pH sensitivity, It is used to the check extent of carbonation in concrete, soap and in toys as diapering work. It is used in Kastle-Meyer test, which identifies the substances containing blood.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phenolphthalein is widely used as an indicator in acid-base titrations, water treatment applications. Due to its pH sensitivity, It is used to the check extent of carbonation in concrete, soap and in toys as diapering work. It is used in Kastle-Meyer test, which identifies the substances containing blood.
Solubility
Soluble in alcohol. Slightly soluble in dimethyl sulfoxide and insoluble in benzene or hexane.
Notes
Acid-base indicator: pH 8.0 - 10.0.Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Acid-base indicator: pH 8.0 - 10.0.
  2. Dobrydneva, Y.; Wilson, E.; Abelt, C. J.; Blackmore, P. F. Phenolphthalein as a Prototype Drug for a Group of Structurally Related Calcium Channel Blockers in Human Platelets. J. Cardiovasc. Pharmacol. 2009, 53 (3), 231-240.
  3. Vishwakarma, N. K.; Patel, V. K.; Hira, S. K.; Ramesh, K.; Srivastava, P.; Mitra, K.; Singh, S.; Chattopadhyay, D.; Maiti, P.; Misra, N.; Manna, P. P.; Ray, B. Tadpole-shaped Beta-cyclodextrin-tagged poly(N-vinylpyrrolidone): synthesis, characterization and studies of its complexation with phenolphthalein and anti tumor activities. RSC Adv. 2015, 5 (20), 15547-15558.