Check Order Status
Diisobutyl phthalate, 99%, Thermo Scientific™
Element Search
Structure Search
Diisobutyl phthalate, 99%, Thermo Scientific™
Have Questions?
Catalog Number
Quantity
ALFA17059.36
500 g
Catalog number
ALFA17059.36
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or Material
Diisobutyl phthalate
CAS
84-69-5
Health Hazard 1
H360Df
Health Hazard 2
GHS H Statement
H360
May damage fertility or the unborn child.
Health Hazard 3
P201-P202-P281-P308+P313-P501c