Sodium salicylate, 99%
Sodium salicylate, 99%
Sodium salicylate, 99%
Sodium salicylate, 99%
Thermo Scientific Chemicals
Sodium salicylate, 99%
CAS: 54-21-7 | C7H5NaO3 | 160.104 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number ALFA17056.30
Price (MYR)
377.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
377.00
EA
Add to cart
Specifications
Chemical Name or MaterialSodium salicylate
CAS54-21-7
Health Hazard 1H302-H319
Health Hazard 2GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
View more
Sodium salicylate is used as an analgesic and antipyretic. It also acts as a non-steroidal anti-inflammatory drug (NSAID). Further, it is used as a phosphor for the detection of vacuum ultraviolet radiation and electrons. It serves as a neurofibromatosis kappa B inhibitor and reducer of oxidative stress. It finds applications as a drug with the function of being antifebrile and anti-rheumatic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium salicylate is used as an analgesic and antipyretic. It also acts as a non-steroidal anti-inflammatory drug (NSAID). Further, it is used as a phosphor for the detection of vacuum ultraviolet radiation and electrons. It serves as a neurofibromatosis kappa B inhibitor and reducer of oxidative stress. It finds applications as a drug with the function of being antifebrile and anti-rheumatic.
Solubility
Soluble in water, 1,4-dioxane, ethanol and glycerol.
Notes
Light sensitive. Store in a cool place. Incompatible with strong oxidizing agents, strong acidsmineral acids, metallic salts and iodine.
RUO – Research Use Only
General References:
  1. Younatham, R.; Siefers, K.; Wegman-Points, L.; Pierce, G. Sodium salicylate prevents TNF alpha-induced reductions in insulin-stimulated eNOSser1177 phosphorylation but not Akt in human aortic endothelial cells. FASEB J. 2015, 29 (1), 802-805.
  2. Li, Q.; Qi, Y.; Gao, C. Chemical regeneration of spent powdered activated carbon used in decolorization of sodium salicylate for the pharmaceutical industry. J. Cleaner Prod. 2015, 86, 424-431.