n-Butyl benzoate, 98%
n-Butyl benzoate, 98%
n-Butyl benzoate, 98%
n-Butyl benzoate, 98%
Thermo Scientific Chemicals
n-Butyl benzoate, 98%
CAS: 136-60-7 | C11H14O2 | 178.23 g/mol
Have Questions?
Catalog NumberQuantity
ALFA17010.30250 g
Catalog number ALFA17010.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or Materialn-Butyl benzoate
CAS136-60-7
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c