D-Lyxose, 99%
D-Lyxose, 99%
D-Lyxose, 99%
Thermo Scientific Chemicals
D-Lyxose, 99%
CAS: 1114-34-7 | C5H10O5 | 150.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
Catalog number ALFA17007.14
Price (MYR)
1,583.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
1,583.00
EA
Add to cart
Specifications
Chemical Name or MaterialD-Lyxose
CAS1114-34-7
Melting Point109°C to 115°C
Recommended StorageAmbient temperatures
Density1.545