Ammonium citrate tribasic, 97+%
Ammonium citrate tribasic, 97+%
Ammonium citrate tribasic, 97+%
Ammonium citrate tribasic, 97+%
Ammonium citrate tribasic, 97+%
Thermo Scientific Chemicals
Ammonium citrate tribasic, 97+%
CAS: 3458-72-8 | C6H20N3O7+3 | 246.24 g/mol
Have Questions?
Catalog NumberQuantity
ALFA16973.22100 g
Catalog number ALFA16973.22
Price (MYR)
392.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
392.00
EA
Add to cart
Specifications
Chemical Name or MaterialAmmonium citrate tribasic
CAS3458-72-8
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Melting Point∼180°C (decomposition)
View more
Lithium nitrate is used in ceramics, pyrotechnics, salt baths, heat-exchange media, refrigeration systems, rocket propellant and in solar batteries. It acts as an oxidizing agent used in the production of red-colored fireworks and flares, as a catalyst to accelerate the breakdown of nitrogen oxides and as a medium to store heat received from sun for cooking. It is applied to concrete-pavement to withstand weathering effects.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Solubility
Soluble in water. Slightly soluble in alcohol.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Gajadeera, C. S.; Zhang, X.; Wei, Y.; Tsodikov, O. V.; Structure of inorganic pyrophosphatase from Staphylococcus aureus reveals conformational flexibility of the active site. J. Struct. Biol. 2015, 189 (2), 81-86.
  2. Xue, L.; Liao, Y.; Yang, L.; Li, X.; Li, W. Improved rate performance of LiNi0.5Mn1.5O4 cathode for lithium ion battery by carbon coating. Ionics 2015, 21 (5), 1269-1275.