4-(Trifluoromethyl)benzyl bromide, 98%
4-(Trifluoromethyl)benzyl bromide, 98%
4-(Trifluoromethyl)benzyl bromide, 98%
4-(Trifluoromethyl)benzyl bromide, 98%
Thermo Scientific Chemicals
4-(Trifluoromethyl)benzyl bromide, 98%
CAS: 402-49-3 | C8H6BrF3 | 239.035 g/mol
Have Questions?
Catalog NumberQuantity
ALFA16895.065 g
Catalog number ALFA16895.06
Price (MYR)
559.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
559.00
EA
Add to cart
Specifications
Chemical Name or Material4-(Trifluoromethyl)benzyl bromide
CAS402-49-3
Health Hazard 1H314
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
View more
4-(Trifluoromethyl)benzyl bromide can be used to produce p-trifluoromethylbenzyl phenyl sulfide. It is used in medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-(Trifluoromethyl)benzyl bromide can be used to produce p-trifluoromethylbenzyl phenyl sulfide. It is used in medicine.
Solubility
Insoluble in water.
Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents. Incompatible with oxidizing agents and bases.
RUO – Research Use Only
M. Karnan; V. Balachandran; M. Murugan; M.K. Murali; A. Nataraj. Vibrational (FT-IR and FT-Raman) spectra, NBO, HOMO-LUMO, Molecular electrostatic potential surface and computational analysis of 4-(trifluoromethyl)benzylbromide. Spectrochimica Acta Part A: Molecular and Biomolecular Spectroscopy.2013, 116 84-95.