Naphthalene-2,6-dicarboxylic acid, 98+%
Naphthalene-2,6-dicarboxylic acid, 98+%
Naphthalene-2,6-dicarboxylic acid, 98+%
Naphthalene-2,6-dicarboxylic acid, 98+%
Thermo Scientific Chemicals
Naphthalene-2,6-dicarboxylic acid, 98+%
CAS: 1141-38-4 | C12H8O4 | 216.192 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFA16868.14
Price (MYR)
980.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
980.00
EA
Add to cart
Specifications
Chemical Name or MaterialNaphthalene-2,6-dicarboxylic acid
CAS1141-38-4
Melting Point>300°C
Recommended StorageAmbient temperatures
EINECS Number214-527-0