3-(Trifluoromethyl)benzoic acid, 98+%
3-(Trifluoromethyl)benzoic acid, 98+%
3-(Trifluoromethyl)benzoic acid, 98+%
3-(Trifluoromethyl)benzoic acid, 98+%
Thermo Scientific Chemicals
3-(Trifluoromethyl)benzoic acid, 98+%
CAS: 454-92-2 | C8H5F3O2 | 190.12 g/mol
Have Questions?
Catalog NumberQuantity
ALFA16737.0910 g
Catalog number ALFA16737.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or Material3-(Trifluoromethyl)benzoic acid
CAS454-92-2
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c