1-Pentene, 97%
1-Pentene, 97%
1-Pentene, 97%
Thermo Scientific Chemicals
1-Pentene, 97%
CAS: 109-67-1 | C5H10 | 70.14 g/mol
Have Questions?
Catalog NumberQuantity
ALFA16723.1425 g
Catalog number ALFA16723.14
Price (MYR)
450.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
450.00
EA
Add to cart
Specifications
Chemical Name or Material1-Pentene
CAS109-67-1
Health Hazard 1H224-H304
Health Hazard 2GHS H Statement
H224-H304
Extremely flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Health Hazard 3P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
View more
1-Pentene is used in the preparation of d-xylitol, l-arabinitol and hydrazones of substituted salicylaldehydes. It is also used as a blending agent for of high-octane gasoline. It reacts with p-toluenesulfinate to yield 1-(2-iodo-pentane-1-sulfonyl)-4-methyl-benzene. It is also involved in Gas Chromatography and Liquid Chromatography analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Pentene is used in the preparation of d-xylitol, l-arabinitol and hydrazones of substituted salicylaldehydes. It is also used as a blending agent for of high-octane gasoline. It reacts with p-toluenesulfinate to yield 1-(2-iodo-pentane-1-sulfonyl)-4-methyl-benzene. It is also involved in Gas Chromatography and Liquid Chromatography analysis.
Solubility
Miscible with ethanol, ethyl ether, benzene and organic solvents. Immiscible with water.
Notes
Incompatible with strong oxidizing agents, reducing agents, halogens and acids.
RUO – Research Use Only
General References:
  1. von Aretin, T.; Schallmoser, S.; Standl, S.; Tonigold, M.; Lercher, J. A.; Hinrichsen, O. Single-Event Kinetic Model for 1-Pentene Cracking on ZSM-5. Ind. Eng. Chem. Res. 2015, 54 (47), 11792-11803.
  2. Peirone, S. A.; Cometto, P. M.; Lane, S. I. OH-Initiated Photooxidations of 1-Pentene and 2-Methyl-2-propen-1-ol: Mechanism and Yields of the Primary Carbonyl Products. ChemPhysChem 2014, 15 (17), 3848-3854.