1,2,3,4-Butanetetracarboxylic acid, 98+%
1,2,3,4-Butanetetracarboxylic acid, 98+%
1,2,3,4-Butanetetracarboxylic acid, 98+%
1,2,3,4-Butanetetracarboxylic acid, 98+%
Thermo Scientific Chemicals
1,2,3,4-Butanetetracarboxylic acid, 98+%
CAS: 1703-58-8 | C8H10O8 | 234.16 g/mol
Have Questions?
Catalog NumberQuantity
ALFA16612.22100 g
Catalog number ALFA16612.22
Price (MYR)
424.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
424.00
EA
Add to cart
Specifications
Chemical Name or Material1,2,3,4-Butanetetracarboxylic acid
CAS1703-58-8
Health Hazard 1H302-H318
Health Hazard 2GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
View more
1,2,3,4-Butanetetracarboxylic acid is used as a cross linking agent to prepare cotton cellulose in order to improve anti-prilling and flame retardant properties. It is used as a spacer in the cross-linking of titania nanoparticles to cotton. Further, it is used as a nonformaldehyde durable press finishing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,2,3,4-Butanetetracarboxylic acid is used as a cross linking agent to prepare cotton cellulose in order to improve anti-prilling and flame retardant properties. It is used as a spacer in the cross-linking of titania nanoparticles to cotton. Further, it is used as a nonformaldehyde durable press finishing agent.
Solubility
Soluble in water and ethanol.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Ji, B.; Tang, P.; Yan, K.; Sun, G. Catalytic actions of alkaline salts in reactions between 1,2,3,4-butanetetracarboxylic acid and cellulose: II. Esterification. Carbohydr. Polym. 2015, 132, 228-236.
  2. Kawase, T.; Nomura, Y.; Oida, T. Synthesis and Properties of 1,2,3,4-Butanetetracarboxylic Acid Type Gemini Surfactants with Semifluoroalkyl Chain as Hydrophobic Group. Tenside Surfact. Det. 2014, 52 (5), 386-395.