D-Pantothenic acid calcium salt, 98%
D-Pantothenic acid calcium salt, 98%
D-Pantothenic acid calcium salt, 98%
D-Pantothenic acid calcium salt, 98%
Thermo Scientific Chemicals
D-Pantothenic acid calcium salt, 98%
CAS: 137-08-6 | C18H34CaN2O11 | 494.551 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFA16609.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialD-Pantothenic acid calcium salt hydrate
Melting Point∼138°C (decomposition)
CAS137-08-6
Recommended StorageAmbient temperatures
RTECS NumberRU4375000
View more
Vitamin B-5D-Pantothenic acid calcium salt hydrate acts as a precursor in the biosynthesis of coenzyme A. It also involved in the catabolism of fatty acids, preparation of heme and the neurotransmitter acetylcholine. It is involved in the regulation of gene expression and in signal transduction. Further, it is used in sphingolipids, cholesterol and steroid hormones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Vitamin B-5D-Pantothenic acid calcium salt hydrate acts as a precursor in the biosynthesis of coenzyme A. It also involved in the catabolism of fatty acids, preparation of heme and the neurotransmitter acetylcholine. It is involved in the regulation of gene expression and in signal transduction. Further, it is used in sphingolipids, cholesterol and steroid hormones.
Solubility
Soluble in water.
Notes
Hygroscopic. Air, light and moisture sensitive. Incompatible with strong oxidizing agents. Store in a cool place. Keep the container tightly closed.
RUO – Research Use Only
Poh, C. K.; Ng, S.; Lim, T. Y.; Tan, H. C.; Loo, J.; Wang, W. In vitro characterizations of mesoporous hydroxyapatite as a controlled release delivery device for VEGF in orthopedic applications. J. Biomed. Mater. Res. Part A 2012, 100A (11), 3143-3150.