n-Nonane, 99%
n-Nonane, 99%
n-Nonane, 99%
Thermo Scientific Chemicals
n-Nonane, 99%
CAS: 111-84-2 | C9H20 | 128.259 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ALFA16177.AP
Price (MYR)
1,232.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
1,232.00
EA
Add to cart
Specifications
Chemical Name or Materialn-Nonane
CAS111-84-2
Health Hazard 1H226-H304-H315-H319-H335-H336
Health Hazard 2GHS H Statement
H304-H226-H332
May be fatal if swallowed and enters airways.
Flammable liquid and vapour.
Harmful if inhaled.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
View more
Nonane is used in organic synthesis, as a solvent, as a distillation chaser and as a fuel additive. It is also used in biodegradable detergent. It is an active ingredient of such petroleum fractions as varnish makers and painter naphtha, 140 flash, stoddard solvents and in gasoline.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Nonane is used in organic synthesis, as a solvent, as a distillation chaser and as a fuel additive. It is also used in biodegradable detergent. It is an active ingredient of such petroleum fractions as varnish makers and painter naphtha, 140 flash, stoddard solvents and in gasoline.
Solubility
Miscible with ethanol, ether, acetone, benzene, chloroform and hydrogen peroxide. Immiscible with water.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Cripwell, J. T.; Schwarz, C. E.; Burger, A. J. Vapor-Liquid Equilibria Measurements for the Nine n-Alkane/Ketone Pairs Comprising 2-, 3-, and 4-Heptanone with n-Octane, n-Nonane, and n-Decane. J. Chem. Eng. Data 2015, 60 (3), 602-611.
  2. Mansfield, E.; Outcalt, S. L. Bubble-Point Measurements of n-Butane+ n-Octane and n-Butane+ n-Nonane Binary Mixtures. J. Chem. Eng. Data 2015, 60 (8), 2447-2453.