Citral, cis + trans, 95%
Citral, cis + trans, 95%
Citral, cis + trans, 95%
Citral, cis + trans, 95%
Thermo Scientific Chemicals
Citral, cis + trans, 95%
CAS: 5392-40-5 | C10H16O | 152.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
50 g
100 g
Catalog number ALFA16169.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCitral
Name Notecis + trans
CAS5392-40-5
Health Hazard 1H315-H317-H319
Health Hazard 2GHS H Statement
H315-H317