Polyethylene glycol 4,000
Polyethylene glycol 4,000
Polyethylene glycol 4,000
Polyethylene glycol 4,000
Thermo Scientific Chemicals
Polyethylene glycol 4,000
CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number ALFA16151.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPolyethylene glycol 4,000
pH5 to 7
CAS25322-68-3
Melting Point58°C to 62°C
Recommended StorageAmbient temperatures
View more
Polyethylene glycol is used in gene delivery of protoplast transfection and incubation. It is used in the preparation of extraction for measuring cytosolic phosphoenolpyruvate carboxykinase (PEPCK) from plant tissues. It is useful to mediate DNA transformation into protoplasts derived from rice leaf sheaths.