Strontium acetate hemihydrate, 98%
Strontium acetate hemihydrate, 98%
Strontium acetate hemihydrate, 98%
Strontium acetate hemihydrate, 98%
Thermo Scientific Chemicals
Strontium acetate hemihydrate, 98%
CAS: 14692-29-6 | Sr(OOCCH3)2·0.5H2O
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFA16149.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialStrontium acetate hemihydrate
CAS14692-29-6
Melting Point150°C (in -0.5H2O)
Recommended StorageAmbient temperatures
EINECS Number208-854-8
View more
PST precursor solutions were prepared from lead (II) acetate,titanium tetraisopropoxide, and strontium acetate hemihydrate with solvents of acetic acid and 2-methoxyethanol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
PST precursor solutions were prepared from lead (II) acetate,titanium tetraisopropoxide, and strontium acetate hemihydrate with solvents of acetic acid and 2-methoxyethanol.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Y. Duan,J. Li, X. Yang, X. -M. Cao, L. Hu, Z. -Y. Wang, Y. -W. Liu , C. -X. Wang.Thermal investigation of strontium acetate hemihydrate in nitrogen gas . Journal of Thermal Analysis and Calorimetry. 2008, 941, 169-174.
  2. Sushil K. Misra,; Krishan Kumar. Single crystal EPR study of Cu2+-doped strontium acetate hemihydrate [Sr(CH3COO)2 · Full-size image (<1 K)H2O] . Physica B+C. 1986, 138 (1-2), 171-175.