Cobalt(II,III) oxide
Cobalt(II,III) oxide
Cobalt(II,III) oxide
Cobalt(II,III) oxide
Thermo Scientific Chemicals
Cobalt(II,III) oxide
CAS: 1308-06-1 | Co3O4 | 240.80 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1000 g
Catalog number ALFA16121.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCobalt(II,III) oxide
Melting Point900°C (decomposition)
CAS1308-06-1
Health Hazard 1H334-H350i
Health Hazard 2GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
View more
Cobalt(II,III) oxide is currently utilized in the process of artificial photosynthesis. It is also used for glass, optic and ceramic applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cobalt(II,III) oxide is currently utilized in the process of artificial photosynthesis. It is also used for glass, optic and ceramic applications.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Xi Wang; Wei Tian; Tianyou Zhai; Chunyi Zhi; Yoshio Bando; Dmitri Golberg. Cobalt(II,III) oxide hollow structures: fabrication, properties and applications. Journal of Materials Chemistry. 2012, 22, (44),23310-23326
  2. Q.Li; T.Sasaki; N.Koshizaki. Pressure dependence of the morphology and size of cobalt (II,III) oxide nanoparticles prepared by pulsed-laser ablation. Applied Physics A. 1999, 69, (1),115-118