4-Chloro-2-fluorobenzaldehyde, 98%
4-Chloro-2-fluorobenzaldehyde, 98%
4-Chloro-2-fluorobenzaldehyde, 98%
4-Chloro-2-fluorobenzaldehyde, 98%
Thermo Scientific Chemicals
4-Chloro-2-fluorobenzaldehyde, 98%
CAS: 61072-56-8 | C7H4ClFO | 158.556 g/mol
Have Questions?
Catalog NumberQuantity
ALFA16110.0910 g
Catalog number ALFA16110.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Chloro-2-fluorobenzaldehyde
CAS61072-56-8
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
4-Chloro-2-fluorobenzaldehyde is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Chloro-2-fluorobenzaldehyde is used as pharmaceutical intermediates.
Solubility
Insoluble in water.
Notes
Air sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only
Bruce C.Hamper; Kindrick L.Leschinsky; Steven S.Massey; Crystal L.Bell; Lawrence H. Brannigan; S.Douglas Prosch. Synthesis and herbicidal activity of 3-aryl-5-(haloalkyl)-4-isoxazolecarboxamides and their derivatives. J. Agric. Food Chem. 1995, 43, (1),219-228