Bis(2-ethylhexyl) sebacate, 95%
Bis(2-ethylhexyl) sebacate, 95%
Bis(2-ethylhexyl) sebacate, 95%
Bis(2-ethylhexyl) sebacate, 95%
Thermo Scientific Chemicals
Bis(2-ethylhexyl) sebacate, 95%
CAS: 122-62-3 | C26H50O4 | 426.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 mL
250 mL
Catalog number ALFA16049.AU
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialBis(2-ethylhexyl) sebacate
CAS122-62-3
Recommended StorageAmbient temperatures
Density0.914
EINECS Number204-558-8
View more
Bis(2-ethylhexyl) sebacate is used as a plasticizer. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Bis(2-ethylhexyl) sebacate is used as a plasticizer. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
Solubility
Soluble in water (0.1 g/L at 20°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Markus Rothmaier, Wilhelm Simon. Chloride-selective electrodes based on mercury organic compounds as neutral carriers. Analytica Chimica Acta. 1993, 271 (1), 135-141.
  2. Yuji Kawabata.; Toshito Kamichika.; Totaro Imasaka.; Nobuhiko Ishibashi. Ion-selective optrode using hexadecyl-Acridine Orange attached on poly(vinyl chloride) membrane. Anal. Chem. 1990, 62 (18), 2054-2055.