4-Chlorochalcone, 98+%
4-Chlorochalcone, 98+%
4-Chlorochalcone, 98+%
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number ALFA15978.18
Price (MYR)
-
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Chlorochalcone
CAS956-04-7
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more