Sodium diethyldithiocarbamate trihydrate, 98%
Sodium diethyldithiocarbamate trihydrate, 98%
Sodium diethyldithiocarbamate trihydrate, 98%
Sodium diethyldithiocarbamate trihydrate, 98%
Thermo Scientific Chemicals
Sodium diethyldithiocarbamate trihydrate, 98%
CAS: 20624-25-3 | C5H16NNaO3S2 | 225.297 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15898.22100 g
Catalog number ALFA15898.22
Price (MYR)
417.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
417.00
EA
Add to cart
Specifications
Chemical Name or MaterialSodium diethyldithiocarbamate trihydrate
CAS20624-25-3
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
Sodium diethyldithiocarbamate trihydrate is a spin trapping reagent used in conjunction with ferrous ion to detect nitric oxide in brain, kidney, liver and other tissues. It is used as sensitive reagent for the determination of copper, rubber vulcanization accelerator and finds application in the determination of other metals such as zinc, cobalt, platinum and palladium. Furthermore, it is an inhibitor of superoxide dismutase, ascorbate oxidase and other enzymes as well as inhibits the induction of macrophage nitric oxide synthase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium diethyldithiocarbamate trihydrate is a spin trapping reagent used in conjunction with ferrous ion to detect nitric oxide in brain, kidney, liver and other tissues. It is used as sensitive reagent for the determination of copper, rubber vulcanization accelerator and finds application in the determination of other metals such as zinc, cobalt, platinum and palladium. Furthermore, it is an inhibitor of superoxide dismutase, ascorbate oxidase and other enzymes as well as inhibits the induction of macrophage nitric oxide synthase.
Solubility
Soluble in water, ethanol, methanol and acetone. Insoluble in ether.
Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Reagent for extraction of heavy metals: Anal. Chem., 54, 2536 (1982).
  2. Pang, H.; Chen, D.; Cui, Q. C.; Dou, Q. P. Sodium diethyldithiocarbamate, an AIDS progression inhibitor and a copper-binding compound, has proteasome-inhibitory and apoptosis-inducing activities in cancer cells. Int. J. Mol. Med. 2007, 19 (5), 809-816.
  3. Radovan, C.; Manea, F. Determination of Sodium Diethyldithiocarbamate in Water by Anodic Voltammetry Using a Boron-Doped Diamond Electrode. Electroanalysis 2007, 19 (1), 91-95.