L-Arginine, 98+%
L-Arginine, 98+%
L-Arginine, 98+%
Thermo Scientific Chemicals
L-Arginine, 98+%
CAS: 74-79-3 | C6H14N4O2 | 174.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFA15738.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialL-Arginine
Melting Point226°C (decomposition)
CAS74-79-3
Health Hazard 1H319
Health Hazard 3P264b-P280i-P305+P351+P338
View more
L-Arginine is used for heart and blood vessel conditions which includes congestive heart failure (CHF), chest pain, high blood pressure and coronary artery disease. It plays a vital role in the treatment of cardiovascular disease due to it being antiatherogenic, anti-ischemic, antiplatelet and antithrombotic. It acts as a growth stimulant and is involved in the treatment of erectile dysfunction in men. It is an important ingredient of tooth paste which provides effective relief for sensitive teeth.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in water. Slightly soluble in ethanol. Insoluble in ethyl ether
Notes
Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Mommersteeg, P. M.; Schoemaker, R. G.; Eisel, U. L.; Garrelds, I. M.; Schalkwijk, C. G.; Kop, W. J. Nitric Oxide Dysregulation in Patients With Heart Failure: The Association of Depressive Symptoms With L-Arginine, Asymmetric Dimethylarginine, Symmetric Dimethylarginine, and Isoprostane. Psychosom. Med. 2015, 77 (3), 292-302.
  2. Schneider, J. Y.; Rothmann, S.; Schröder, F.; Langen, J.; Lucke, T.; Mariotti, F.; Huneau, J. F.; Frolich, J. C.; Tsikas, D. Effects of chronic oral l-arginine administration on the l-arginineO pathway in patients with peripheral arterial occlusive disease or coronary artery disease: l-Arginine prevents renal loss of nitrite, the major NO reservoir. Amino Acids 2015, 47 (9), 1961-1974.