Ellagic acid hydrate, 97%, may cont. up to 12% water
Ellagic acid hydrate, 97%, may cont. up to 12% water
Ellagic acid hydrate, 97%, may cont. up to 12% water
Ellagic acid hydrate, 97%, may cont. up to 12% water
Thermo Scientific Chemicals
Ellagic acid hydrate, 97%, may cont. up to 12% water
CAS: 476-66-4 | C14H6O8 | 302.19 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15722.065 g
Catalog number ALFA15722.06
Price (MYR)
396.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
396.00
EA
Add to cart
Specifications
Chemical Name or MaterialEllagic acid hydrate
CAS476-66-4
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Ellagic acid used as selective, ATP-competitive inhibitor of casein kinase 2. Exhibits antitumor activity and inhibits glutathione S-transferase. It is also used as a Topo I and II, FGR, GSK, and PKA inhibitor.Commonly occurring plant polyphenol, inhibitor of glutathione S-transferase. Used for the assay of factor XIIa in plasma.Contact activation in blood coagulation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ellagic acid used as selective, ATP-competitive inhibitor of casein kinase 2. Exhibits antitumor activity and inhibits glutathione S-transferase. It is also used as a Topo I and II, FGR, GSK, and PKA inhibitor.Commonly occurring plant polyphenol, inhibitor of glutathione S-transferase. Used for the assay of factor XIIa in plasma.Contact activation in blood coagulation.
Solubility
Slightly soluble in water. Soluble in 1M sodium hydroxide (10 mg/ml), EtOH, and pyridine.
Notes
Air sensitive and light sensitive. Store under dry inert gas. Store in cool dry place in tightly closed container. Store in dark and away form air.
RUO – Research Use Only
General References:
  1. Patrice Njomnang Soh: Benoît Witkowski; David Olagnier; Marie-Laure Nicolau; Maria-Concepcion Garcia-Alvarez; Antoine Berry; and Françoise Benoit-Vical. In Vitro and In Vivo Properties of Ellagic Acid in Malaria Treatment. Antimicrob. Agents Chemother. 2009, 31100-1106.
  2. Sungwoo Kim; Satoru K. Nishimoto; Joel D. Bumgardnerd; Warren O. Haggardd; M. Waleed Gabere; Yunzhi Yang. A chitosan/β-glycerophosphate thermo-sensitive gel for the delivery of ellagic acid for the treatment of brain cancer. Biomaterials. 2010, 31 (14), 4157-4166.