p-Phenylenediamine, 97%
p-Phenylenediamine, 97%
p-Phenylenediamine, 97%
p-Phenylenediamine, 97%
Thermo Scientific Chemicals
p-Phenylenediamine, 97%
CAS: 106-50-3 | C6H8N2 | 108.144 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number ALFA15680.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or Materialp-Phenylenediamine
CAS106-50-3
Health Hazard 1H301+H311+H331-H317-H319
Health Hazard 2GHS H Statement
H301-H311-H331-H319-H317
Health Hazard 3P261-P264b-P270-P271-P272-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P333+P313-P361-P363-P501c
View more
p-Phenylenediamine is used as a precursor to aramid plastics, Kevlar fibers and to urethane polymers. It serves as a developing agent in photography, antioxidants in rubber products and as a vulcanization accelerator. It is used as a henna surrogate for temporary tattoos and as a histological stain for lipids such as myelin. It is an anti-fade reagent used in fluorescence microscopy to retard photobleaching of fluorescein and other fluorescent dyes. Furthermore, it is also an ingredient in hair dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
p-Phenylenediamine is used as a precursor to aramid plastics, Kevlar fibers and to urethane polymers. It serves as a developing agent in photography, antioxidants in rubber products and as a vulcanization accelerator. It is used as a henna surrogate for temporary tattoos and as a histological stain for lipids such as myelin. It is an anti-fade reagent used in fluorescence microscopy to retard photobleaching of fluorescein and other fluorescent dyes. Furthermore, it is also an ingredient in hair dyes.
Solubility
Soluble in alcohol, chloroform, ether and hot benzene.
Notes
Store in cool place. Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Magdziarz, P.; Bober, P.; Trchová, M.; Morávková, Z.; Bláha, M., Prokeš, J.; Stejskal, J. Conducting composites prepared by the reduction of silver ions with poly (p-phenylenediamine). Polym. Int. 2015, 64 (4), 496-504.
  2. Pot, L. M.; Coenraads, P. J.; Goebel, C.; Blömeke, B. Assessment of the elicitation response in subjects weakly sensitized to p-phenylenediamine. Brit. J. Dermatol. 2015, 172 (1), 138-144.